5-Chloro-1H-pyrazolo[4,3-b]pyridine
Leverantör: Apollo Scientific
Varning
OR301099-1G OR301099-5G
APOSOR301099-1GEA 790 SEK
APOSOR301099-1G APOSOR301099-5G
5-Chloro-1H-pyrazolo[4,3-b]pyridine
5-Chloro-1H-pyrazolo[4,3-b]pyridine
Formel: C₆H₄ClN₃
MW: 153,5691 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD05663984
CAS nummer: 94220-45-8