1-Bromo-2-chloro-3-nitrobenzene 98%
Leverantör: Apollo Scientific
|
Varning
|
OR301062-1G
OR301062-5G
OR301062-10G
APOSOR301062-1GEA
775
SEK
APOSOR301062-1G
APOSOR301062-5G
APOSOR301062-10G
1-Bromo-2-chloro-3-nitrobenzene 98%
1-Bromo-2-chloro-3-nitrobenzene
Formel:
C₆H₃BrClNO₂ MW: 236,4505 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD08277301 CAS nummer: 3970-37-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...