1H-Pyrrolo[2,3-b]pyridin-5-ol 97%
Leverantör: Apollo Scientific
Varning
Synonyms: 5-Hydroxy-7-azaindole
OR301023-25G OR301023-5G OR301023-1G
APOSOR301023-25GEA 2225 SEK
APOSOR301023-25G APOSOR301023-5G APOSOR301023-1G
1H-Pyrrolo[2,3-b]pyridin-5-ol 97%
1H-Pyrrolo[2,3-b]pyridin-5-ol
Formel: C₇H₆N₂O
MW: 134,14 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD08272242
CAS nummer: 98549-88-3