tert-Butyl-4-(3-hydroxypropyl)tetrahydro-1(2H)-pyridinecarboxylate

Leverantör: Apollo Scientific
Varning

Synonyms: tert-Butyl 4-(3-hydroxypropyl)piperidine-1-carboxylate, 4-(3-Hydroxyprop-1-yl)piperidine, N-BOC protected, tert-Butyl 4-(3-hydroxypropyl)tetrahydro-1(2H)-pyridinecarboxylate

OR30070-1G OR30070-250MG
APOSOR30070-1GEA 1075 SEK
APOSOR30070-1G APOSOR30070-250MG
tert-Butyl-4-(3-hydroxypropyl)tetrahydro-1(2H)-pyridinecarboxylate
tert-Butyl-4-(3-hydroxypropyl)tetrahydro-1(2H)-pyridinecarboxylate
Formel: C₁₃H₂₅NO₃
MW: 243,35 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD02677712
CAS nummer: 156185-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR