3,5-Dibromopyridin-4-ol
Leverantör: Apollo Scientific
Varning
Synonyms: 3,5-Dibromo-4-hydroxypyridine
OR300639-1G OR300639-5G
APOSOR300639-1GEA 720 SEK
APOSOR300639-1G APOSOR300639-5G
3,5-Dibromopyridin-4-ol
3,5-Dibromopyridin-4-ol
Formel: C5H3Br2NO
MW: 252,8914 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00082969
CAS nummer: 25813-25-6