5-Oxopyrrolidine-3-carboxylic acid methyl ester

Leverantör: Apollo Scientific
Varning

Synonyms: Methyl 5-oxopyrrolidine-3-carboxylate

OR300522-5G OR300522-25G
APOSOR300522-5GEA 1025 SEK
APOSOR300522-5G APOSOR300522-25G
5-Oxopyrrolidine-3-carboxylic acid methyl ester
5-Oxopyrrolidine-3-carboxylic acid methyl ester
Formel: C₆H₉NO₃
MW: 143,14 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD08275678
CAS nummer: 35309-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR