6-Chloro-1H-pyrazolo[4,3-b]pyridine
Leverantör: Apollo Scientific
Varning
OR300409-1G OR300409-250MG
APOSOR300409-1GEA 2650 SEK
APOSOR300409-1G APOSOR300409-250MG
6-Chloro-1H-pyrazolo[4,3-b]pyridine
6-Chloro-1H-pyrazolo[4,3-b]pyridine
Formel: C₆H₄ClN₃
Storage Temperature: Omgivande
MDL Number: MFCD16659575
CAS nummer: 1260670-01-6