4-Bromo-1H-pyrazolo[4,3-c]pyridine
Leverantör: Apollo Scientific
Varning
OR300386-1G OR300386-250MG OR300386-100MG
APOSOR300386-1GEA 1850 SEK
APOSOR300386-1G APOSOR300386-250MG APOSOR300386-100MG
4-Bromo-1H-pyrazolo[4,3-c]pyridine
4-Bromo-1H-pyrazolo[4,3-c]pyridine
Formel: C₆H₄BrN₃
MW: 198.03 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD12401001
CAS nummer: 1159829-63-6