5-Chloro-2-hydroxynicotinic acid 97%
Leverantör: Apollo Scientific
Varning
Synonyms: 5-Chloro-2-hydroxy-3-pyridinecarboxylic Acid
OR29878-25G OR29878-5G
APOSOR29878-25GEA 1400 SEK
APOSOR29878-25G APOSOR29878-5G
5-Chloro-2-hydroxynicotinic acid 97%
5-Chloro-2-hydroxynicotinic acid
Formel: C₆H₄ClNO₃
MW: 173,55 g/mol
Smältpunkt: 258…260 °C
Storage Temperature: Omgivande
MDL Number: MFCD00204158
CAS nummer: 38076-80-1