2-Bromo-5-iodopyrazine 98%

Leverantör: Apollo Scientific
Varning

OR2986-1G OR2986-5G
APOSOR2986-1GEA 260 SEK
APOSOR2986-1G APOSOR2986-5G
2-Bromo-5-iodopyrazine 98%
2-Bromo-5-iodopyrazine
Formel: C₄H₂BrIN₂
Smältpunkt: 95…97 °C
Storage Temperature: Kylskåp
MDL Number: MFCD08061602
CAS nummer: 622392-04-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR