2-Chloroisonicotinonitrile
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
2-Chloro-4-cyanopyridine, 2-Chloropyridine-4-carbonitrile
OR29832-10G
OR29832-250G
OR29832-50G
OR29832-500G
APOSOR29832-10GEA
175
SEK
APOSOR29832-10G
APOSOR29832-250G
APOSOR29832-50G
APOSOR29832-500G
2-Chloroisonicotinonitrile
2-Chloroisonicotinonitrile
Formel:
C₆H₃ClN₂ MW: 138,56 g/mol Kokpunkt: 104…106 °C/15mmHg Smältpunkt: 69…73 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00174318 CAS nummer: 33252-30-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...