4-Bromo-1-ethyl-3-methyl-1H-pyrazole-5-carboxylic acid
Leverantör: Apollo Scientific
Varning
OR29671-1G OR29671-250MG
APOSOR29671-1GEA 1300 SEK
APOSOR29671-1G APOSOR29671-250MG
4-Bromo-1-ethyl-3-methyl-1H-pyrazole-5-carboxylic acid
4-Bromo-1-ethyl-3-methyl-1H-pyrazole-5-carboxylic acid
Formel: C₇H₉BrN₂O₂
MW: 233.626 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00173776
CAS nummer: 175276-99-0