3-Aminoisonicotinic acid 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 3-Aminopyridine-4-carboxylic acid
OR29468-25G OR29468-5G
APOSOR29468-25GEA 1200 SEK
APOSOR29468-25G APOSOR29468-5G
3-Aminoisonicotinic acid 98%
3-Aminoisonicotinic acid
Formel: C₆H₆N₂O₂
MW: 138,13 g/mol
Smältpunkt: 287…291 °C
Storage Temperature: Omgivande
MDL Number: MFCD00137840
CAS nummer: 7579-20-6