Pyridine-3,4-dicarboxylic acid
Leverantör: Apollo Scientific
Varning
Synonyms: 3,4-Pyridinedicarboxylic acid, Cinchomeronic acid, Pyridine-3,4-dicarboxylicacid
OR28966-100G OR28966-25G OR28966-500G
APOSOR28966-100GEA 620 SEK
APOSOR28966-100G APOSOR28966-25G APOSOR28966-500G
Pyridine-3,4-dicarboxylic acid
Pyridine-3,4-dicarboxylic acid
Formel: C₇H₅NO₄
MW: 167,12 g/mol
Smältpunkt: 262 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00006392
CAS nummer: 490-11-9
EINECS: 207-705-4