3-Bromophenyl isocyanate 97%
Leverantör: Apollo Scientific
Synonyms: 1-Bromo-3-isocyanatobenzene
OR28901-25G OR28901-5G
APOSOR28901-25GEA 3800 SEK
APOSOR28901-25G APOSOR28901-5G
3-Bromophenyl isocyanate 97%
3-Bromophenyl isocyanate
  • May form small amount of sediment.
Formel: C₇H₄BrNO
MW: 198,02 g/mol
Kokpunkt: 220 °C
Densitet: 1,586 g/cm³ (25 °C)
Flampunkt: 108 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00002014
CAS nummer: 23138-55-8