N-Aminophthalimide 98%

Leverantör: Apollo Scientific

Synonyms: 2-Aminoisoindoline-1,3-dione

OR28752-100G OR28752-25G OR28752-5G
APOSOR28752-100GEA 4975 SEK
APOSOR28752-100G APOSOR28752-25G APOSOR28752-5G
N-Aminophthalimide 98%
N-Aminophthalimide
Formel: C₈H₆N₂O₂
MW: 162,15 g/mol
Smältpunkt: 200…202 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005895
CAS nummer: 1875-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR