2-Bromoanisole 98%
Leverantör: Apollo Scientific
|
|
Synonyms:
1-Bromo-2-methoxybenzene
OR28661-100G
OR28661-25G
APOSOR28661-100GEA
295
SEK
APOSOR28661-100G
APOSOR28661-25G
2-Bromoanisole 98%
2-Bromoanisole
Formel:
C₇H₇BrO MW: 187,04 g/mol Kokpunkt: 220…223 °C Smältpunkt: 44257 °C Densitet: 1,502 g/cm³ (25 °C) Flampunkt: 96 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00000064 CAS nummer: 578-57-4 EINECS: 209-425-8 UN: 3082 ADR: 9+EHS,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...