2-Chloromethyl-m-xylene
Leverantör: Apollo Scientific
Fara
Synonyms: α-Chloro-1,2,6-trimethylbenzene, 1-(Chloromethyl)-2,6-dimethylbenzene, 2,6-Dimethylbenzyl chloride, 2-(Chloromethyl)-1,3-dimethylbenzene
OR2853-25G OR2853-5G
APOSOR2853-25GEA 275 SEK
APOSOR2853-25G APOSOR2853-5G
2-Chloromethyl-m-xylene
2-Chloromethyl-m-xylene
Formel: C₉H₁₁Cl
MW: 154,64 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD01310815
CAS nummer: 5402-60-8
UN: 3261
ADR: 8,II