5-Aminoisophthalic acid

Leverantör: Apollo Scientific
Varning

OR28141-1KG OR28141-500G
APOSOR28141-1KGEA 2100 SEK
APOSOR28141-1KG APOSOR28141-500G
5-Aminoisophthalic acid
5-Aminoisophthalic acid
Formel: C₈H₇NO₄
MW: 181,15 g/mol
Smältpunkt: >300 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002522
CAS nummer: 99-31-0
EINECS: 202-748-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR