Methyl-3-methyl-2-nitrobenzoate 98%

Leverantör: Apollo Scientific
Varning

Synonyms: Methyl 3-methyl-2-nitrobenzoate

OR27752-100G OR27752-25G OR27752-500G
APOSOR27752-100GEA 1425 SEK
APOSOR27752-100G APOSOR27752-25G APOSOR27752-500G
Methyl-3-methyl-2-nitrobenzoate 98%
Methyl-3-methyl-2-nitrobenzoate
Formel: C₉H₉NO₄
MW: 195,17 g/mol
Smältpunkt: 72…76 °C
Storage Temperature: Omgivande
MDL Number: MFCD01003947
CAS nummer: 5471-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR