4-(2,5-Dimethyl-1H-pyrrol-1-yl)benzenecarboxylic acid

Leverantör: Apollo Scientific
Varning

Synonyms: 4-(2,5-Dimethyl-1H-pyrrol-1-yl)benzoic acid

OR27723-10G OR27723-1G OR27723-5G
APOSOR27723-10GEA 5400 SEK
APOSOR27723-10G APOSOR27723-1G APOSOR27723-5G
4-(2,5-Dimethyl-1H-pyrrol-1-yl)benzenecarboxylic acid
4-(2,5-Dimethyl-1H-pyrrol-1-yl)benzenecarboxylic acid
Formel: C₁₃H₁₃NO₂
MW: 215,25 g/mol
Smältpunkt: 181…184 °C
Storage Temperature: Omgivande
MDL Number: MFCD00453921
CAS nummer: 15898-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR