2-Chloro-6-methylisonicotinic acid

Leverantör: Apollo Scientific
Varning

Synonyms: 2-Chloro-6-methylpyridine-4-carboxylic acid

OR27369-10G OR27369-25G OR27369-5G
APOSOR27369-10GEA 875 SEK
APOSOR27369-10G APOSOR27369-25G APOSOR27369-5G
2-Chloro-6-methylisonicotinic acid
2-Chloro-6-methylisonicotinic acid
Formel: C₇H₆ClNO₂
MW: 171,58 g/mol
Smältpunkt: 200 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00052830
CAS nummer: 25462-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR