Cyclopropanecarboxamidine hydrochloride
Leverantör: Apollo Scientific
Varning
OR27201-25G OR27201-5G OR27201-100G
APOSOR27201-25GEA 380 SEK
APOSOR27201-25G APOSOR27201-5G APOSOR27201-100G
Cyclopropanecarboxamidine hydrochloride
Cyclopropanecarboxamidine hydrochloride
Formel: C₄H₈N₂
Smältpunkt: 122…127 °C
Storage Temperature: Omgivande
MDL Number: MFCD00053010
CAS nummer: 57297-29-7