2,6-Dichloro-3-cyano-4-picoline 97%

Leverantör: Apollo Scientific
Fara

Synonyms: 2,6-Dichloro-4-methylnicotinonitrile, 2,6-dichloro-4-methylpyridine-3-carbonitrile, 2,6-Dichloro-3-cyano-4-methylpyridine

OR27041-100G OR27041-25G OR27041-250G
APOSOR27041-100GEA 1400 SEK
APOSOR27041-100G APOSOR27041-25G APOSOR27041-250G
2,6-Dichloro-3-cyano-4-picoline 97%
2,6-Dichloro-3-cyano-4-picoline
Formel: C₇H₄Cl₂N₂
MW: 187,03 g/mol
Smältpunkt: 108…112 °C
Storage Temperature: Omgivande
MDL Number: MFCD00052633
CAS nummer: 875-35-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR