5-Chloroanthranilic acid 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 2-Amino-5-chlorobenzoic acid
OR26820-100G OR26820-25G OR26820-500G
APOSOR26820-100GEA 225 SEK
APOSOR26820-100G APOSOR26820-25G APOSOR26820-500G
5-Chloroanthranilic acid 98%
5-Chloroanthranilic acid
Formel: C₇H₆ClNO₂
MW: 171,58 g/mol
Smältpunkt: 208…213 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00007838
CAS nummer: 635-21-2
EINECS: 211-230-8