6-Quinolineboronic acid
Leverantör: Apollo Scientific
Varning
Synonyms: Quinolin-6-ylboronic acid, Quinoline-6-boronic acid
OR2621-250MG OR2621-1G OR2621-5G
APOSOR2621-250MGEA 225 SEK
APOSOR2621-250MG APOSOR2621-1G APOSOR2621-5G
6-Quinolineboronic acid
6-Quinolineboronic acid
Formel: C₉H₈BNO₂
MW: 172,98 g/mol
Smältpunkt: 106…109 °C
Storage Temperature: Kylskåp
MDL Number: MFCD03093100
CAS nummer: 376581-24-7