2-Methyl-4-pyridineboronic acid

Leverantör: Apollo Scientific
Varning

Synonyms: (2-Methylpyridin-4-yl)boronic acid, 2-Methylpyridine-4-boronic acid

OR2618-1G OR2618-5G
APOSOR2618-1GEA 260 SEK
APOSOR2618-1G APOSOR2618-5G
2-Methyl-4-pyridineboronic acid
2-Methyl-4-pyridineboronic acid
Formel: C₆H₈BNO₂
MW: 136,9442 g/mol
Smältpunkt: 122…123 °C
Storage Temperature: Frys
MDL Number: MFCD03095364
CAS nummer: 579476-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR