(2-Iodophenyl)acetic acid
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
2-(2-Iodophenyl)acetic acid
OR26135-10G
OR26135-5G
APOSOR26135-10GEA
895
SEK
APOSOR26135-10G
APOSOR26135-5G
(2-Iodophenyl)acetic acid
(2-Iodophenyl)acetic acid
Formel:
(C₆H₄I)CH₂COOH MW: 262,05 g/mol Smältpunkt: 115…117 °C Storage Temperature: Kylskåp |
MDL Number:
MFCD00046546 CAS nummer: 18698-96-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...