2-Phenylisonicotinic acid 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 2-Phenylpyridine-4-carboxylic acid, 2-Phenylisonicotinic acid
OR2592-1G OR2592-250MG
APOSOR2592-1GEA 1500 SEK
APOSOR2592-1G APOSOR2592-250MG
2-Phenylisonicotinic acid 98%
2-Phenylisonicotinic acid
Formel: C₁₂H₉NO₂
Smältpunkt: 266…268 °C
Storage Temperature: Omgivande
MDL Number: MFCD03086008
CAS nummer: 55240-51-2