2-(Methylsulphonyl)benzoic acid 98%

Leverantör: Apollo Scientific
Varning

OR2589-25G OR2589-5G
APOSOR2589-25GEA 6825 SEK
APOSOR2589-25G APOSOR2589-5G
2-(Methylsulphonyl)benzoic acid 98%
2-(Methylsulphonyl)benzoic acid
Formel: C₈H₈O₄S
MW: 200,21 g/mol
Smältpunkt: 137…140 °C
Storage Temperature: Omgivande
MDL Number: MFCD00216492
CAS nummer: 33963-55-2
EINECS: 223-765-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR