Isoxazole 98%

Leverantör: Apollo Scientific
Fara

OR2580-1G OR2580-25G OR2580-5G
APOSOR2580-1GEA 175 SEK
APOSOR2580-1G APOSOR2580-25G APOSOR2580-5G
Isoxazole 98%
Isoxazole
Formel: C₃H₃NO
MW: 69,06 g/mol
Kokpunkt: 93…95 °C
Densitet: 1,078 g/cm³ (25 °C)
Flampunkt: 12 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003149
CAS nummer: 288-14-2
UN: 2924
ADR: 3+8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR