N-(tert-Butyl)-2-chloroacetamide
Leverantör: Apollo Scientific
|
Varning
|
OR25511-1G
OR25511-5G
APOSOR25511-1GEA
670
SEK
APOSOR25511-1G
APOSOR25511-5G
N-(tert-Butyl)-2-chloroacetamide
N-(tert-Butyl)-2-chloroacetamide
Formel:
C₆H₁₂ClNO MW: 149,62 g/mol Smältpunkt: 79…82 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00032565 CAS nummer: 15678-99-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...