4-Oxotetrahydrothiophene-3-carboxylic acid methyl ester, Technical Grade
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
Methyl 4-oxotetrahydrothiophene-3-carboxylate
OR25037-10G
OR25037-1G
OR25037-250MG
APOSOR25037-10GEA
1125
SEK
APOSOR25037-10G
APOSOR25037-1G
APOSOR25037-250MG
4-Oxotetrahydrothiophene-3-carboxylic acid methyl ester, Technical Grade
4-Oxotetrahydrothiophene-3-carboxylic acid methyl ester
Formel:
C₆H₈O₃S MW: 160,19 g/mol Kokpunkt: 80…81 °C/0,5mmHg Storage Temperature: Kylskåp |
MDL Number:
MFCD00052381 CAS nummer: 2689-68-1 EINECS: 220-256-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...