4-Methyl-2,3-dihydro-1H-pyrrolo[3,4-c]quinoline-1,3-dione

Leverantör: Apollo Scientific
Varning

OR25028-1G OR25028-250MG OR25028-500MG OR25028-5G
APOSOR25028-1GEA 2575 SEK
APOSOR25028-1G APOSOR25028-250MG APOSOR25028-500MG APOSOR25028-5G
4-Methyl-2,3-dihydro-1H-pyrrolo[3,4-c]quinoline-1,3-dione
4-Methyl-2,3-dihydro-1H-pyrrolo[3,4-c]quinoline-1,3-dione
Formel: C₁₂H₈N₂O₂
MW: 212,21 g/mol
Smältpunkt: 255…256 °C
Storage Temperature: Omgivande
MDL Number: MFCD00179682
CAS nummer: 27295-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR