4-(4-Acetylpiperazin-1-yl)benzoic acid
Leverantör: Apollo Scientific
OR2498-1G OR2498-250MG OR2498-500MG OR2498-2.5G OR2498-5G
APOSOR2498-1GEA 4625 SEK
APOSOR2498-1G APOSOR2498-250MG APOSOR2498-500MG APOSOR2498-2.5G APOSOR2498-5G
4-(4-Acetylpiperazin-1-yl)benzoic acid
4-(4-Acetylpiperazin-1-yl)benzoic acid
Formel: C₁₃H₁₆N₂O₃
MW: 248.28 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD05862631
CAS nummer: 104080-55-9