α-Bromo-p-toluic acid 98%
Leverantör: Apollo Scientific
Fara
Synonyms: 4-Bromomethylbenzoic acid, 4-(Bromomethyl)benzoic acid
OR24888-100G OR24888-25G
APOSOR24888-100GEA 1550 SEK
APOSOR24888-100G APOSOR24888-25G
α-Bromo-p-toluic acid 98%
α-Bromo-p-toluic acid
Formel: C₈H₇BrO₂
MW: 215,05 g/mol
Smältpunkt: 224…229 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002567
CAS nummer: 6232-88-8
UN: 3261
ADR: 8,III