N-Benzyl-N-(2-chloroethyl)amine 90+%
Leverantör: Apollo Scientific
Varning
OR24839-1G OR24839-250MG OR24839-100MG
APOSOR24839-1GEA 2925 SEK
APOSOR24839-1G APOSOR24839-250MG APOSOR24839-100MG
N-Benzyl-N-(2-chloroethyl)amine 90+%
N-Benzyl-N-(2-chloroethyl)amine
Formel: C₉H₁₂ClN
MW: 169,66 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD01005550
CAS nummer: 42074-16-8