3-Hexylthiophene 98%

Leverantör: Apollo Scientific
Varning

OR24741-25G OR24741-100G
APOSOR24741-25GEA 1350 SEK
APOSOR24741-25G APOSOR24741-100G
3-Hexylthiophene 98%
3-Hexylthiophene
Formel: C₁₀H₁₆S
MW: 168,3 g/mol
Kokpunkt: 65 °C/0,45mmHg
Densitet: 0,936 g/cm³ (25 °C)
Flampunkt: 37 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00143181
CAS nummer: 1693-86-3
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR