2,2,5,5-Tetramethyl-3-pyrrolin-1-oxyl-3-carboxylic acid free radical
Leverantör: Apollo Scientific
Varning
Synonyms: 1-Hydroxy-2,2,5,5-tetramethyl-2,5-dihydro-1H-pyrrole-3-carboxylic acid, 1H-Pyrrol-1-yloxy, 3-carboxy-2,5-dihydro-2,2,5,5-tetramethyl-, (3-Carboxy-2,2,5,5-tetramethyl-2,5-dihydro-1H-pyrrol-1-yl)oxidany
OR24671-1G OR24671-250MG
APOSOR24671-1GEA 6600 SEK
APOSOR24671-1G APOSOR24671-250MG
2,2,5,5-Tetramethyl-3-pyrrolin-1-oxyl-3-carboxylic acid free radical
2,2,5,5-Tetramethyl-3-pyrrolin-1-oxyl-3-carboxylic acid
Formel: C₉H₁₄NO₃
MW: 184.212 g/mol
Smältpunkt: 202 °C
Storage Temperature: Omgivande
MDL Number: MFCD00041847
CAS nummer: 2154-67-8