9-(Chloromethyl)anthracene 97%
Leverantör: Apollo Scientific
|
Fara
|
OR24592-25G
OR24592-5G
APOSOR24592-25GEA
1825
SEK
APOSOR24592-25G
APOSOR24592-5G
9-(Chloromethyl)anthracene 97%
9-(Chloromethyl)anthracene
Formel:
C₁₅H₁₁Cl MW: 226,71 g/mol Smältpunkt: 138…142 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00001263 CAS nummer: 24463-19-2 EINECS: 246-274-7 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...