O-tert-Butylhydroxylamine hydrochloride 98%
Leverantör: Apollo Scientific
Varning
Synonyms: O-(tert-Butyl)hydroxylamine hydrochloride
OR24583-25G OR24583-5G
APOSOR24583-25GEA 2225 SEK
APOSOR24583-25G APOSOR24583-5G
O-tert-Butylhydroxylamine hydrochloride 98%
O-tert-Butylhydroxylamine hydrochloride
Formel: C₄H₁₂ClNO
MW: 125,5972 g/mol
Smältpunkt: 155 °C
Storage Temperature: Omgivande
MDL Number: MFCD00043272
CAS nummer: 39684-28-1
EINECS: 254-590-1
UN: 1325
ADR: 4.1,III