N-Acetylguanidine
Leverantör: Apollo Scientific
|
Varning
|
OR24582-25G
OR24582-5G
OR24582-10G
APOSOR24582-25GEA
345
SEK
APOSOR24582-25G
APOSOR24582-5G
APOSOR24582-10G
N-Acetylguanidine
N-Acetylguanidine
Formel:
C₃H₇N₃O Smältpunkt: 181…183 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00075615 CAS nummer: 5699-40-1 EINECS: 227-180-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...