Phenylboronic acid 99%
Leverantör: Apollo Scientific
Varning
Synonyms: Phenylmetaboric acid, Dihydroxy(phenyl)borane, Phenyl boric acid, Benzeneboronic acid
OR2447-100G OR2447-25G
APOSOR2447-100GEA 650 SEK
APOSOR2447-100G APOSOR2447-25G
Phenylboronic acid 99%
Phenylboronic acid
Formel: C₆H₇BO₂
MW: 121.93 g/mol
Smältpunkt: 216…219 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00002103
CAS nummer: 98-80-6
EINECS: 202-701-9
Merck Index: 13,01068