2-Chloro-4-(dimethylamino)benzaldehyde 95+%

Leverantör: Apollo Scientific
Varning

OR2435-10G OR2435-5G
APOSOR2435-10GEA 2625 SEK
APOSOR2435-10G APOSOR2435-5G
2-Chloro-4-(dimethylamino)benzaldehyde 95+%
2-Chloro-4-(dimethylamino)benzaldehyde
Formel: C₉H₁₀ClNO
MW: 183,64 g/mol
Smältpunkt: 83…84 °C
Storage Temperature: Omgivande
MDL Number: MFCD00021051
CAS nummer: 1424-66-4
EINECS: 215-840-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR