Aluminon
Leverantör: Apollo Scientific
Varning
Synonyms: Ammonium 5,5'-((3-carboxylato-4-oxocyclohexa-2,5-dien-1-ylidene)methylene)bis(2-hydroxybenzoate), Aurin Tricarboxylic Acid Ammonium Salt
OR2429-100G OR2429-250G
APOSOR2429-100GEA 225 SEK
APOSOR2429-100G APOSOR2429-250G
Aluminon
Aluminon
Formel: C₂₂H₂₃N₃O₉
MW: 473,44 g/mol
Smältpunkt: 220…225 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00040925
CAS nummer: 569-58-4
EINECS: 209-319-1