Cyanimidodithiocarbonic acidmonomethyl ester monopotassium salt 95%
Leverantör: Apollo Scientific
Varning
Synonyms: Potassium methyl N-cyanocarbonodithioimidate
OR24285-10G OR24285-1G OR24285-5G
APOSOR24285-10GEA 6700 SEK
APOSOR24285-10G APOSOR24285-1G APOSOR24285-5G
Cyanimidodithiocarbonic acidmonomethyl ester monopotassium salt 95%
Cyanimidodithiocarbonic acidmonomethyl ester monopotassium salt
Formel: C₃H₃KN₂S₂
MW: 170,289 g/mol
Smältpunkt: 210…212 °C
Storage Temperature: Omgivande
MDL Number: MFCD00799296
CAS nummer: 10191-61-4