3,3,5-Trimethylheptane
Leverantör: Apollo Scientific
|
Fara
|
OR2380-1ML
OR2380-5ML
APOSOR2380-1MLEA
5075
SEK
APOSOR2380-1ML
APOSOR2380-5ML
3,3,5-Trimethylheptane
3,3,5-Trimethylheptane
Formel:
C₁₀H₂₂ Storage Temperature: Omgivande |
CAS nummer:
7154-80-5 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...