3-(2,6-Dichlorophenoxy)-1-propyne
Leverantör: Apollo Scientific
Synonyms: 1,3-Dichloro-2-(prop-2-ynyloxy)benzene
OR23682-10G OR23682-1G OR23682-5G
APOSOR23682-10GEA 3850 SEK
APOSOR23682-10G APOSOR23682-1G APOSOR23682-5G
3-(2,6-Dichlorophenoxy)-1-propyne
3-(2,6-Dichlorophenoxy)-1-propyne
Formel: C₉H₆Cl₂O
MW: 201,05 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00052717
CAS nummer: 3598-66-1