[3-Methyl-5-phenyl-4-isoxazolyl]methanol

Leverantör: Apollo Scientific
Varning

OR23340-10G OR23340-1G OR23340-5G
APOSOR23340-10GEA 3850 SEK
APOSOR23340-10G APOSOR23340-1G APOSOR23340-5G
[3-Methyl-5-phenyl-4-isoxazolyl]methanol
[3-Methyl-5-phenyl-4-isoxazolyl]methanol
Formel: C₁₁H₁₁NO₂
Smältpunkt: 110…112 °C
Storage Temperature: Omgivande
CAS nummer: 113826-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR