4-(4-Methylpiperazin-1-yl)benzoic acid
Leverantör: Apollo Scientific
Varning
Synonyms: 1-(4-Carboxyphenyl)-4-methylpiperazine, 4-(4-Methylpiperazinyl)benzoic acid
OR23308-1G OR23308-250MG OR23308-5G
APOSOR23308-1GEA 295 SEK
APOSOR23308-1G APOSOR23308-250MG APOSOR23308-5G
4-(4-Methylpiperazin-1-yl)benzoic acid
4-(4-Methylpiperazin-1-yl)benzoic acid
Formel: C₁₂H₁₆N₂O₂
MW: 220.2700 g/mol
Smältpunkt: 270…272 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD02682063
CAS nummer: 86620-62-4